missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ERp57/PDIA3 Recombinant Protein Antigen

Código de producto. 18617305 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
18617305 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18617305

Marca: Novus Biologicals™ NBP236766PEP

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Recombinant Protein

TRUSTED_SUSTAINABILITY

Specifications

Para utilizar con (aplicación) Antibody Competition
Formulación PBS and 1 M Urea, pH 7.4.
ID de gen (Entrez) 2923
Peso molecular MolecularWeight-theroretical: 31 kDa
Cantidad 100 μL
Requisitos de almacenamiento Store at -20°C. Avoid freeze/thaw cycles.
Alias de gen Disulfide isomerase ER-60, EC 5.3.4.1, endoplasmic reticulum P58, Endoplasmic reticulum resident protein 57, Endoplasmic reticulum resident protein 60, ER protein 60, ER60, ERP57, ERp5758 kDa glucose-regulated protein, ERP60, ERp6058 kDa microsomal protein, ERp61, glucose regulated protein, 58 kDa, GRP57, GRP58ER protein 57, HsT17083, P58, phospholipase C-alpha, PI-PLC, protein disulfide isomerase family A, member 3, protein disulfide isomerase-associated 3, protein disulfide-isomerase A3
Símbolo de gen PDIA3
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Propiedades del inhibidor This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-36766. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/mL
Formulario Purified
Categoría de investigación Cellular Markers, ER Markers, Membrane Trafficking and Chaperones
Grado de pureza o calidad >80% by SDS-PAGE and Coomassie blue staining
Expresión ERp57/PDIA3
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.