Learn More
1-(2,4-Difluorofenil)piperazina, 99 %, Thermo Scientific™
24.20€ - 314.00€
Identificadores de productos químicos
CAS | 115761-79-0 |
---|---|
Fórmula molecular | C10H12F2N2 |
Peso molecular (g/mol) | 198.217 |
Número MDL | MFCD00082588 |
Clave InChI | CMCSPBOWEYUGHB-UHFFFAOYSA-N |
PubChem CID | 2734637 |
Nombre IUPAC | 1-(2,4-difluorofenil)piperazina |
SMILES | C1CN(CCN1)C2=C(C=C(C=C2)F)F |
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Identificadores de productos químicos
115761-79-0 | |
198.217 | |
CMCSPBOWEYUGHB-UHFFFAOYSA-N | |
1-(2,4-difluorofenil)piperazina |
C10H12F2N2 | |
MFCD00082588 | |
2734637 | |
C1CN(CCN1)C2=C(C=C(C=C2)F)F |
Especificaciones
1-(2,4-Difluorophenyl)piperazine | |
74°C to 76°C | |
C10H12F2N2 | |
UN2923 | |
Air Sensitive | |
CMCSPBOWEYUGHB-UHFFFAOYSA-N | |
1-(2,4-difluorofenil)piperazina | |
2734637 | |
99% |
115761-79-0 | |
108°C to 111°C (0.2 mmHg) | |
MFCD00082588 | |
8620369 | |
1-2,4-difluorophenyl piperazine, 1-2,4-difluorophenyl-piperazine, piperazine, 1-2,4-difluorophenyl, 2,4-difluorophenyl piperazine, pubchem9352, acmc-1ca8h, 2,4-difluorophenylpiperazine, 4-2,4-difluorophenyl piperazine, 1-2,4-difluoro-phenyl-piperazine, piperazine,1-2,4-difluorophenyl | |
C1CN(CCN1)C2=C(C=C(C=C2)F)F | |
198.217 | |
198.22 |
Seguridad y manipulación
GHS H Statement
H301-H314-H318
Toxic if swallowed.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Temperaturas ambientes
RUO – Research Use Only