Learn More
ZWILCH Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14281-1-AP-20UL
This item is not returnable.
View return policy
Description
Essential component of the mitotic checkpoint, which prevents cells from prematurely exiting mitosis. Required for the assembly of the dynein-dynactin and MAD1-MAD2 complexes onto kinetochores.Specifications
| ZWILCH | |
| Polyclonal | |
| Unconjugated | |
| ZWILCH | |
| hZwilch, KNTC1AP, Protein zwilch homolog, ZWILCH | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55055, 68014, 691493 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q8R060, Q9H900 | |
| ZWILCH | |
| ZWILCH Fusion Protein Ag5454 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |