Description
Essential component of the mitotic checkpoint, which prevents cells from prematurely exiting mitosis. Required for the assembly of the dynein-dynactin and MAD1-MAD2 complexes onto kinetochores.
Specifications
Specifications
| Antígeno | ZWILCH |
| Aplicaciones | Immunohistochemistry (Paraffin), Western Blot |
| Clasificación | Polyclonal |
| Concentración | 0.27 mg/mL |
| Conjugado | Unconjugated |
| Formulación | PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 |
| génica | ZWILCH |
| N.º de referencia del gen | Q8R060, Q9H900 |
| Alias de gen | hZwilch, KNTC1AP, Protein zwilch homolog, ZWILCH |
| Símbolos de los genes | ZWILCH |
| Show More |