Learn More
Abnova™ ZNF2 Recombinant Protein
Product Code. 16171942
Change view
Click to view available options
Cantidad:
10 μg
25 μg
Unit Size:
10 microgramos
25 microgramos
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
16171942 25 μg 25 microgramos
16161942 10 μg 10 microgramos
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16171942 Supplier Abnova™ Supplier No. H00007549P01.25ug
This item is not returnable. View return policy
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Número de acceso AAH05068
Para utilizar con (aplicación) Antibody Production, Protein Array, ELISA, Western Blot
Formulación 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
ID de gen (Entrez) 7549
Peso molecular 57.86
Nombre ZNF2 (Human) Recombinant Protein (P01)
Intervalo de pH 8
Método de preparación In vitro wheat germ expression system
Método de purificación Glutathione Sepharose 4 Fast Flow
Pruebas de control de calidad 12.5% SDS-PAGE Stained with Coomassie Blue.
Cantidad 25 μg
Fuente Wheat Germ (in vitro)
Inmunógeno MGTEKQSPSGETRKKSLSRDKGLRRRSALSREILTKERHQECSDCGKTFFDHSSLTRHQRTHTGEKPYDCRECGKAFSHRSSLSRHLMSHTGESPYECSVCSKAFFDRSSLTVHQRIHTGEKPFQCNECGKAFFDRSSLTRHQRIHTGESPYECHQCGKAFSQKSILTRHQLIHTGRKPYECNECGKAFYGVSSLNRHQKAHAGDPRYQCNECGKAFFDRSSLTQHQKIHTGDKPHECSECGKAFSQRCRLTRHQRVHTGEKPFECTVCGKVFSSKSSVIQHQRRYAKQGID
Requisitos de almacenamiento Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Alias de gen A1-5/ZNF661/Zfp661
Nombre común ZNF2
Símbolo de gen ZNF2
Reactividad cruzada Human
Especie Wheat Germ (in vitro)
Recombinante Recombinant
Etiqueta de proteína GST
Formulario Solution