missing translation for 'onlineSavingsMsg'
Learn More

Novus Biologicals™ ZNF14 Recombinant Protein Antigen

Código de producto. 18033509 Tienda Bio Techne Productos
Click to view available options
Cantidad:
0,1 mL
Tamaño de la unidad:
0.10 ml
Este artículo no se puede devolver. Vea la política de devoluciones

Product Code. 18033509

missing translation for 'mfr': Novus Biologicals™ NBP213552PEP

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Highly purified. Generating reliable and reproducible results. Applications: Antibody Competition

A recombinant protein antigen with a N-terminal His6-ABP tag corresponding to human ZNF14. The ZNF14 Recombinant Protein Antigen is derived from E. coli. The ZNF14 Recombinant Protein Antigen has been validated for the following applications: Antibody Competition.

This is a blocking peptide for NBP2-13552. This is a human recombinant protein expressed in E. coli with a N-terminal His-ABP tag and purified with IMAC chromatography.

TRUSTED_SUSTAINABILITY

Specifications

Gene ID (Entrez) 7561
Especie Human
Método de purificación Chromatography
Pureza >80%
Concentración 0.5mg/mL
Contenido y almacenamiento Store at -20°C. Avoid freeze-thaw cycles.
Formulación PBS and 1M Urea, pH 7.4.
Para utilizar con (aplicación) Blocking/Neutralizing, Control
Símbolo de gen ZNF14
Tipo de etiqueta Unlabeled
Peso molecular 21kDa
Tipo de producto ZNF14
Cantidad 0,1 mL
Estado normativo RUO
Fuente E.Coli
Reactividad específica This recombinant antigen is only intended to be used as a blocking agent to confirm antibody specificity with the corresponding antibody, catalog number NBP2-13552. It is purified by IMAC chromatography, and the expected concentration is greater than 0.5 mg/ml
Inmunógeno CESRRGSKCGETTSQMPNVNINKETFTGA
Show More Show Less

Para uso exclusivo en investigación

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.