missing translation for 'onlineSavingsMsg'
Learn More

ZBTB40 Antibody, Novus Biologicals™

Código de producto. 18283540 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
100 μL
Tamaño de la unidad:
100 microlitros
Código de producto. Cantidad unitSize
18283540 100 μL 100 microlitros
1 options
Este artículo no se puede devolver. Vea la política de devoluciones

Product Code. 18283540

Brand: Novus Biologicals NBP106564

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZBTB40 Polyclonal specifically detects ZBTB40 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno ZBTB40
Aplicaciones Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 1.0 mg/mL
Conjugado Unconjugated
Dilución Western Blot 1:2000-1:10000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 2-5 mcg/mg lysate, Immunohistochemistry-Paraffin 1:100-1:500
Formulación Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
N.º de referencia del gen NP_055685.3
Alias de gen KIAA0478ZNF923, MGC133098, zinc finger and BTB domain containing 40, zinc finger and BTB domain-containing protein 40
Símbolos de los genes ZBTB40
Especie del huésped Rabbit
Inmunógeno The immunogen recognized by this antibody maps to a region between residue 1140 and 1190 of human zinc finger and BTB domain containing 40 using the numbering given in entry NP_055685.3 (GeneID 9923).
Método de purificación Affinity Purified
Cantidad 100 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 9923
Especies diana Human
Contenido y almacenamiento Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.