missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Z-DNA Antibody, Novus Biologicals™
Sheep Polyclonal Antibody has been used in 2 publications
Marca: Novus Biologicals NB100-749
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Z-DNA Polyclonal specifically detects Z-DNA in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin.
Especificaciones
| Z-DNA | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.5) with No Preservative | |
| Immunogen was brominated poly-(DG-DC)-left handed DNA (Z-DNA) complexed to methylated BSA | |
| 0.125 mg | |
| DNA Repair, DNA replication Transcription Translation and Splicing | |
| Human | |
| Purified |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| 2.1 mg/mL | |
| Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence, Immunoprecipitation 8 ug/ml, Immunohistochemistry-Paraffin 1:200 | |
| Sheep | |
| Protein G purified | |
| RUO | |
| Primary | |
| Store at -20C. Avoid freeze-thaw cycles. | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido