Learn More
YTHDC1 Mouse anti-Human, Unconjugated, Clone: 1F5G11, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 67911-1-IG-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The YTHDC1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human YTHDC1. This antibody recognizes human antigen. The YTHDC1 antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
| YTHDC1 | |
| Monoclonal | |
| Unconjugated | |
| 20 μL | |
| 91746 | |
| IgG1 |
| Western Blot, ELISA | |
| 1F5G11 | |
| Mouse | |
| Primary | |
| Human |