Learn More
Thermo Scientific™ Human YES1, GST Tag Recombinant Protein
Descripción
For maximum recovery please spin prior to use. Aliquoting the 5 μg, 10 μg, and 20 μg sizes of kinase is not recommended as materials can be used in original packaging until exhausted. For larger sizes, the number of freeze/thaws may be reduced by preparing aliquots; aliquots below 20 μL are not recommended. Never store a kinase diluted. If properly stored at -80C, this product is guaranteed for 6 months from date of purchase. Active-this protein can be used in kinase activity assays, such as Z'-LYTE™ assays, or in a binding assay platform like the LanthaScreen™ Eu Kinase Binding Assay (KBA). Purified-purity is determined by gel electrophoresis and provided in lot-specific certificate of analysis (COA). GST-tagged-provides animal origin flexibility. When used in the KBA platform, it can be paired with a goat- or rabbit-derived Eu-anti-GST antibody. Protein Form: Full Length.
Especificaciones
Especificaciones
| Número de acceso | P07947 |
| Concentración | See Label |
| Para utilizar con (aplicación) | Kinase Assay |
| Formulación | 50mM tris with 50% glycerol, 0.02% Triton X-100, 150mM NaCl, 2mM DTT, 0.5mM EDTA and no preservative; pH 7.5 |
| ID de gen (Entrez) | 7525 |
| Peso molecular | 88.3 kDa |
| Nombre | Human YES1, GST Tag |
| Intervalo de pH | 7.5 |
| Método de purificación | Purified |
| Cantidad | 10 μg |
| Mostrar más |