missing translation for 'onlineSavingsMsg'
Learn More

XBP1 Antibody - BSA Free, Novus Biologicals™

Código de producto. 18703324 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mg
Tamaño de la unidad:
0.03 miligramo
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18703324 0.025 mg 0.03 miligramo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18703324 Proveedor Novus Biologicals N.º de proveedor NBP1772530.025mg

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody has been used in 3 publications

XBP1 Polyclonal specifically detects XBP1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Specifications

Antígeno XBP1
Aplicaciones Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración 1 mg/mL
Conjugado Unconjugated
Dilución Western Blot 0.5-2 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 5 μg/mL, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 5 μg/mL, Immunofluorescence 20 μg/mL
Formulación PBS with 0.02% Sodium Azide
N.º de referencia del gen BAB82982
Alias de gen TREB5, X-box binding protein 1, X-box-binding protein 1, XBP-1, XBP2Tax-responsive element-binding protein 5
Símbolos de los genes XBP1
Especie del huésped Rabbit
Inmunógeno Antibody was raised against an 18 amino acid synthetic peptide from near the amino terminus of human XBP-1. The immunogen is located within amino acids 40 - 90 of XBP-1. Amino Acid Squence: GLPQARKRQRLTHLSPE
Método de purificación Affinity Purified
Cantidad 0.025 mg
Estado normativo RUO
Disciplina de investigación Cancer, Lipid and Metabolism, Unfolded Protein Response
Primario o secundario Primary
ID de gen (Entrez) 7494
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.