missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
XBP1 Monoclonal antibody specifically detects XBP1 in Human samples. It is validated for Flow Cytometry (Intracellular Staining),CyTOF
Specifications
Specifications
| Antígeno | XBP1 |
| Aplicaciones | Flow Cytometry (Intracellular Staining), CyTOF |
| Clasificación | Monoclonal |
| Clon | 525904 |
| Conjugado | Janelia Fluor 635 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | TREB5, X-box binding protein 1, X-box-binding protein 1, XBP-1, XBP2Tax-responsive element-binding protein 5 |
| Especie del huésped | Mouse |
| Inmunógeno | E. coli-derived recombinant human XBP1S, Ser188-Leu287, Accession # NP_001073007 |
| Cantidad | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?