Learn More
WNT8A Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 30518-1-AP-150UL
Descripción
The WNT8A antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human WNT8A. This antibody recognizes human, rat antigen. The WNT8A antibody has been validated for the following applications: IF, WB, ELISA analysis.
Especificaciones
| WNT8A | |
| 1000 μg/mL | |
| Rabbit | |
| Primary | |
| Human, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, Immunofluorescence, ELISA | |
| Protein Wnt 8a, Protein Wnt 8d, WNT8A, WNT8D | |
| 150 μL | |
| 7478 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |