missing translation for 'onlineSavingsMsg'
Learn More

Wnt-5b Antibody, Biotin, Novus Biologicals™

Código de producto. 18613167 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
18613167 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 18613167 Proveedor Novus Biologicals N.º de proveedor NBP241306B

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody

Wnt-5b Polyclonal antibody specifically detects Wnt-5b in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno Wnt-5b
Aplicaciones Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Conjugado Biotin
Formulación PBS
Alias de gen MGC2648, protein Wnt-5b, wingless-type MMTV integration site family, member 5B, WNT-5B protein
Especie del huésped Rabbit
Inmunógeno Antibody was raised against a 15 amino acid peptide near the center of human WNT5B. The immunogen is located within amino acids 130 - 180 of WNT5. Amino Acid Squence: CSRTARPKDLPRDWL
Método de purificación Peptide affinity purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Signal Transduction, Stem Cell Signaling Pathway, Wnt Signaling Pathway
Primario o secundario Primary
ID de gen (Entrez) 81029
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4°C in the dark.
Formulario Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.