Descripción
Features Benefits:
- Lightweight, stainless-steel tool and rubber grip
- Rubber grip prevents tool from slipping when used with disinfectants
- Lightweight to clean tall windows and hard to reach areas
- Maneuvers easily to use vertically and horizontally
Compatibility:
- Compatible with all Contec's QuickConnect handles making windows easier to reach.
- All components of the squeegee are compatible with common cleanroom cleaners and disinfectants
Press Down Handle Clip
- Allows for easy installation preventing the rubber blade from moving during use
Screw-free channel
- Blades to be replaced quickly and easily saving time
Autoclavable
- Squeegee can be autoclaved at 121°C for 30 minutes
- Ideal for use in all areas of the cleanroom
Especificaciones
Especificaciones
| Tipo de producto | Single-blade Squeegee |
| Material del mango | Acero inoxidable |
| Handle Type | Ergonomic |
| Esterilizables en autoclave | Esterilizable en autoclave |
| Material | Caucho |