missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDR93 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Marca: Novus Biologicals NBP2-31871
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
WDR93 Polyclonal specifically detects WDR93 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Especificaciones
| WDR93 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Q6P2C0 | |
| WDR93 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: PQLTSNPKKKVRQPQLNSLGPISADPLEMDANVSFKGDIKLSLPVYIMKIKPPKPVTGTTFKSPLEVFAKIKDCYGLGSGQNHFIKDSQWEQQAEIFNASY | |
| 0.1 mL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| WD repeat domain 93, WD repeat-containing protein 93 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 56964 | |
| Human | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido