missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
VPS37D Polyclonal specifically detects VPS37D in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Especificaciones
Especificaciones
| Antígeno | VPS37D |
| Aplicaciones | Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:20 - 1:50 |
| Formulación | PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide |
| Alias de gen | vacuolar protein sorting 37 homolog D (S. cerevisiae), williams-Beuren syndrome chromosomal region 24 protein, williams-Beuren syndrome region protein 24 |
| Símbolos de los genes | VPS37D |
| Especie del huésped | Rabbit |
| Inmunógeno | This antibody was developed against Recombinant Protein corresponding to amino acids:LRDLLQDEPKLDRIVRLSRKFQGLQLEREACLASNYALAKENLALRPRLEMGRAALAIKYQELREVAENCADKLQRLEESMHRWSPHC |
| Vis mere |
For Research Use Only
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?