Descripción
VPS26A Monoclonal antibody specifically detects VPS26A in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),KnockDown,Immunocytochemistry/ Immunofluorescence
Especificaciones
Especificaciones
| Antígeno | VPS26A |
| Aplicaciones | Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), KnockDown, Immunocytochemistry/Immunofluorescence |
| Clasificación | Monoclonal |
| Clon | CL2287 |
| Conjugado | DyLight 755 |
| Formulación | 50mM Sodium Borate |
| Alias de gen | FLJ12930, HB58, Hbeta58, hVPS26, PEP8A, vacuolar protein sorting 26 (yeast homolog), vacuolar protein sorting 26 homolog A (S. pombe), vacuolar protein sorting 26 homolog A (yeast), vacuolar protein sorting-associated protein 26A, Vesicle protein sorting 26A, VPS26vacuolar protein sorting 26 (yeast) |
| Especie del huésped | Mouse |
| Inmunógeno | This antibody was developed using a recombinant protein derived from O75436, with the exact immunogen sequence remaining proprietary. |
| Método de purificación | Protein A purified |
| Mostrar más |