missing translation for 'onlineSavingsMsg'
Learn More

Vorapaxar, MedChemExpress

Product Code. 16443298
Encompass_Preferred
Change view
Click to view available options
Cantidad:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10 miligramos
25 miligramos
5 miligramos
50 miligramos
Product Code. Cantidad unitSize
16443298 10 mg 10 miligramos
16476017 5 mg 5 miligramos
16439836 50 mg 50 miligramos
16482706 25 mg 25 miligramos
4 options
This item is not returnable. View return policy

Product Code. 16443298

Brand: Medchem Express HY10119

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

MedChemExpress Vorapaxar (SCH 530348), an antiplatelet agent, is a selective, orally active, and competitive thrombin receptor protease-activated receptor (PAR-1) antagonist (Ki=8.1 nM). Vorapaxar (SCH 530348) inhibits thrombin receptor-activating peptide (TRAP)-induced platelet aggregation in a dose-dependent manner.

Vorapaxar (SCH 530348), an antiplatelet agent, is a selective, orally active, and competitive thrombin receptor protease-activated receptor (PAR-1) antagonist (Ki=8.1 nM). Vorapaxar (SCH 530348) inhibits thrombin receptor-activating peptide (TRAP)-induced platelet aggregation in a dose-dependent manner.

Specifications

Nombre del producto químico o material Vorapaxar
CAS 618385-01-6
Color Off-White
Notas de grado de pureza Research
Fórmula molecular C29H33FN2O4
Cantidad 10 mg
Sinónimo SCH 530348
Información de solubilidad DMSO : 25 mg/mL (50.75 mM; Need ultrasonic)
SMILES O=C(O[C@@H]1C)[C@@]2([H])[C@]1([H])[C@@H](/C=C/C3=NC=C(C=C3)C4=CC=CC(F)=C4)[C@]([C@](C5)([H])C2)([H])CC[C@H]5NC(OCC)=O
Peso molecular (g/mol) 492.58
Formula Weight (peso de la fórmula) 492.58
Porcentaje de pureza 98.72%
Grado Research
Forma física Solid
Para utilizar con (aplicación) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.