Learn More
Novus Biologicals™ Von Willebrand Factor Antibody (INSERM patent anti-vWF) - VHH-Fc, Novus Biologicals™
Human Monoclonal Antibody
Marca: Novus Biologicals™ NBP3-28369-100ug
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Von Willebrand Factor Monoclonal antibody specifically detects Von Willebrand Factor in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Especificaciones
| Von Willebrand Factor | |
| Monoclonal | |
| Unconjugated | |
| P04275 | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex. | |
| Store at -20°C in powder form. Store at -80°C once reconstituted. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| INSERM patent anti-vWF | |
| Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) | |
| coagulation factor VIII VWF, F8, F8VWF, von Willebrand factor, VWD, vWF | |
| vWF | |
| 100 μg | |
| Cancer | |
| 7450 | |
| Human | |
| Purified |