Learn More
Invitrogen™ Human Vitamin D3 Receptor, Ligand Binding Domain, GST Tag Recombinant Protein
Código de producto. 10382363
Change view
Click to view available options
Cantidad:
50 μg
Tamaño de la unidad:
cada uno
10382363 50 μg cada uno
1 options
Código de producto. 10382363 Proveedor Invitrogen™ N.º de proveedor PV4552
Recombinant Protein
Vitamin D3 Receptor, Ligand Binding Domain (VDR-LBD) is a 65.50 kDa recombinant human protein (amino acids 93-427) expressed as a GST-tagged fusion protein in baculovirus-infected insect cells. VDR-LBD was expressed and purified in the absence of exogenous ligands, making it ideal for ligand binding and coregulator displacement studies. Protein Form: Recombinant, Ligand Binding Domain
Steroid receptors are ligand-dependent intracellular proteins that stimulate transcription of specific genes by binding to specific DNA sequences following activation by the appropriate hormone. The 1,25-dihydroxy-vitamin D3 receptor (VDR) belongs to the superfamily of steroid hormone receptors which includes estrogen, progesterone, glucocorticoid, androgen, and thyroid hormone receptors. Vitamin D modulates calcium and phosphorus homeostasis, bone remodeling, cell growth regulation, and differentiation. Studies have found VDR in the intestine, bone, kidney, epidermis, and cells of the endocrine immune system. Repression of T-cell proliferation and interleukin-2 (IL-2) gene expression, for example, occurs when VDR binds within the IL-2 enhancer. Formation of VDR/retinoic X receptor (RXR) heterodimers in the presence of intracellular 1,25(OH)2D3 has been shown to interfere with assembly of nuclear factor of activated T-cells (NFATp)/Fos/Jun/DNA complex and subsequent IL-2 gene transcription. The corresponding gene for the vitamin D receptor is NR1I1.
TRUSTED_SUSTAINABILITY
Especificaciones
Número de acceso P11473
Concentración See Label
Para utilizar con (aplicación) Ligand Binding Assay
Formulación proprietary buffer with no preservative
ID de gen (Entrez) 7421
Nombre Human Vitamin D3 Receptor, Ligand Binding Domain, GST Tag
Cantidad 50 μg
Estado normativo RUO - research use only
Alias de gen 1,25-dihydroxyvitamin D3 receptor; HGNC:12679; I79_025795; Nr1i1; Nuclear receptor subfamily 1 group I member 1; PPP1R163; protein phosphatase 1, regulatory subunit 163; vdr; vdr.S; vdr-A; vitamin D (1,25- dihydroxyvitamin D3) receptor; vitamin D (1,25- dihydroxyvitamin D3) receptor S homeolog; vitamin D (1,25-dihydroxyvitamin D3) receptor; vitamin D nuclear receptor variant 1; vitamin D receptor; vitamin D receptor protein; vitamin D receptor S homeolog; Vitamin D3 receptor; XELAEV_18015993mg; xVDR
Nombre común VDR
Longitud de la proteína 93-427
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Etiqueta de proteína GST-tag
Contenido y almacenamiento -80°C, Avoid Freeze/Thaw Cycles
Sistema de expresión Baculovirus
Formulario Liquid
Subtipo de proteína Hormone Receptors