missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Viral MCK-2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Marca: R&D Systems AF798-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MCK-2 Polyclonal specifically detects MCK-2 in Viral samples. It is validated for Western Blot.
Especificaciones
| MCK-2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| E. coli-derived recombinant viral MCK-2 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Detects recombinant viral MCK-2 in direct ELISAs and Western blots. | |
| Virus | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido