missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Viral HIV-1 Gag p24 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Marca: R&D Systems MAB73603-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
HIV-1 Gag p24 Monoclonal antibody specifically detects HIV-1 Gag p24 in Virus samples. It is validated for Western Blot
Especificaciones
| HIV-1 Gag p24 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 155030 | |
| Virus | |
| Purified |
| Western Blot | |
| 1042904 | |
| Western Blot 1 ug/mL | |
| Q77YG1 | |
| E. coli-derived recombinant viral HIV-1 Gag p24, Pro149-Gly354, Accession # Q77YG1 | |
| 25 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido