Learn More
Vimentin Antibody (CL0157) [Allophycocyanin], Novus Biologicals™
Código de producto. 30290284 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
30290284 0.1 mL 0.10 ml
1 options
Código de producto. 30290284 Proveedor Novus Biologicals N.º de proveedor NBP252867APC
This item is not returnable. View return policy
Mouse Monoclonal Antibody
Vimentin Monoclonal antibody specifically detects Vimentin in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin),Gene Knock-Out
TRUSTED_SUSTAINABILITY
Specifications
Antígeno Vimentin
Aplicaciones Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin), Gene Knock-Out
Clasificación Monoclonal
Clon CL0157
Conjugado APC
Formulación PBS
Alias de gen epididymis secretory sperm binding protein, FLJ36605, VIM, vimentin
Especie del huésped Mouse
Inmunógeno This antibody was developed using a recombinant protein derived from P08670, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Cancer, Cell Biology, Cellular Markers, Growth and Development, Hypoxia, Neuronal Cell Markers, Neuroscience, Signal Transduction, Stem Cell Markers, Stem Cells
Primario o secundario Primary
ID de gen (Entrez) 7431
Especies diana Human
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG1