Learn More
Invitrogen™ Human TMEM168 (aa 457-531) Control Fragment Recombinant Protein
Descripción
Highest antigen sequence indentity to the following orthologs: Mouse (95%), Rat (95%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-139951 (PA5-139951. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.
Especificaciones
Especificaciones
| Número de acceso | Q9H0V1 |
| Concentración | ≥5.0 mg/mL |
| Para utilizar con (aplicación) | Blocking Assay, Control |
| Formulación | 1 M urea, PBS with no preservative; pH 7.4 |
| ID de gen (Entrez) | 64418 |
| Nombre | Human TMEM168 (aa 457-531) Control Fragment |
| Cantidad | 100 μL |
| Estado normativo | RUO |
| Alias de gen | 5730526F17Rik; 8430437G11Rik; AI462344; AI504145; RGD1307778; Tmem168; Transmembrane protein 168 |
| Nombre común | TMEM168 |
| Mostrar más |