Learn More
R&D Systems™ Recombinant Human IL-21 (HEK293 Expressed) Protein
Código de producto. 18786255
Click to view available options
Cantidad:
100 μg
10 μg
1 mg
250 μg
50 μg
Tamaño de la unidad:
1 miligramo
10 microgramos
100 microgramos
250 microgramos
50 microgramos
Código de producto. 18786255
Marca: R&D Systems™ 11393IL100
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Carrier Free Recombinant Protein
Measured in a cell proliferation assay using B9 mouse hybridoma cells. The ED50 for this effect is 3.00-30.0 ng/mL.
TRUSTED_SUSTAINABILITY
Especificaciones
Número de acceso Q9HBE4.3
Para utilizar con (aplicación) Bioactivity
Formulación Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose.
ID de gen (Entrez) 59067
Peso molecular MolecularWeight-observed: 15-17 kDa, under reducing conditions., MolecularWeight-theroretical: 15 kDa
Cantidad 100 μg
Fuente Human embryonic kidney cell, HEK293-derived human IL-21 protein Gln32-Ser162
Requisitos de almacenamiento Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20°C to -70°C as supplied. 1 month, 2°C to 8°C under sterile conditions after reconstitution. 3 months, -20°C to -70°C under sterile conditions after reconstitution.
Concentración de endotoxinas <0.10 EU / 1 μg of the protein by the LAL method.
Alias de gen CVID11, IL21, IL-21 Za11interleukin-21, interleukin 21, interleukin-21 isoform, Za11
Conjugado Unconjugated
Reconstitución Reconstitute 10 μg size at 100 μg/mL and other sizes (at 500 μg/mL in PBS.)
Grado de pureza o calidad >95%, by SDS-PAGE visualized with Silver Staining and quantitative densitometry by Coomassie™ Blue Staining.
Expresión IL-21