Descripción
p53 Monoclonal antibody specifically detects p53 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, ChIP assay (ChIP)
Especificaciones
Especificaciones
| Antígeno | p53 |
| Aplicaciones | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunoprecipitation, ChIP Assay |
| Clasificación | Monoclonal |
| Clon | Pab DO-1 |
| Conjugado | mFluor Violet 610 SE |
| Dilución | Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP) |
| Formulación | 50mM Sodium Borate |
| Alias de gen | Antigen NY-CO-13, FLJ92943, LFS1TRP53, p53, p53 tumor suppressor, P53cellular tumor antigen p53, Phosphoprotein p53, transformation-related protein 53, tumor protein p53, Tumor suppressor p53 |
| Especie del huésped | Mouse |
| Inmunógeno | Recombinant human wild type p53 protein expressed in E.coli |
| Mostrar más |