Descripción
Torsin A Monoclonal antibody specifically detects Torsin A in Human samples. It is validated for Western Blot, Flow Cytometry
Especificaciones
Especificaciones
| Antígeno | Torsin A |
| Aplicaciones | Western Blot, Flow Cytometry |
| Clasificación | Monoclonal |
| Clon | SR1592 |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:500-1:2000, Flow Cytometry 1:50 |
| Formulación | PBS, pH 7.4, 150mM NaCl, 50% glycerol. |
| Alias de gen | DQ2Torsin family 1 member A, Dystonia 1 protein, dystonia 1, torsion (autosomal dominant; torsin A), DYT1torsin-1A, torsin family 1, member A (torsin A) |
| Especie del huésped | Rabbit |
| Inmunógeno | A synthesized peptide derived from human Torsin A (Uniprot #: O14656) |
| Mostrar más |