Descripción
IgD Monoclonal antibody specifically detects IgD in Human samples. It is validated for Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | IgD |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | IGHD/2730R |
| Concentración | 0.2 mg/ml |
| Conjugado | Unconjugated |
| Dilución | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Formulación | 1mM PBS with 0.05% BSA |
| Alias de gen | IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta |
| Especie del huésped | Rabbit |
| Mostrar más |
Título del producto
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?