Descripción
BMI-1 Monoclonal antibody specifically detects BMI-1 in Human, Mouse samples. It is validated for Western Blot
Especificaciones
Especificaciones
| Antígeno | BMI-1 |
| Aplicaciones | Western Blot |
| Clasificación | Monoclonal |
| Clon | S08-7A6 |
| Concentración | 0.3 mg/mL |
| Conjugado | Unconjugated |
| Formulación | 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA |
| Alias de gen | B lymphoma Mo-MLV insertion region 1 homolog, B lymphoma Mo-MLV insertion region 1 homolog (mouse), BMI1 polycomb ring finger oncogene, FLVI2/BMI1, MGC12685, murine leukemia viral (bmi-1) oncogene homolog, PCGF4, polycomb complex protein BMI-1, polycomb group protein Bmi1, polycomb group ring finger 4, RING finger protein 51, RNF51Polycomb group RING finger protein 4 |
| Especie del huésped | Rabbit |
| Inmunógeno | Recombinant protein of human BMI-1 (Uniprot # P35226) |
| Mostrar más |