Learn More
SCAPER Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 16994-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CCNA2/CDK2 regulatory protein that transiently maintains CCNA2 in the cytoplasm.Especificaciones
| SCAPER | |
| Polyclonal | |
| Unconjugated | |
| SCAPER | |
| KIAA1454, MSTP063, SCAPER, Zfp291, Zinc finger protein 291, ZNF291 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 117521, 244891, 49855 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9BY12 | |
| SCAPER | |
| SCAPER Fusion Protein Ag10679 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |