Learn More
enQuireBio™ Recombinant other Shiga Like Toxin 2 Protein
Código de producto. 15913759
Change view
Click to view available options
Cantidad:
100 μg
1 mg
500 μg
Tamaño de la unidad:
1 miligramo
100 microgramos
500 microgramos
Código de producto. Cantidad unitSize
15913759 100 μg 100 microgramos
15933759 500 μg 500 microgramos
15923759 1 mg 1 miligramo
3 options
Código de producto. 15913759
Marca: enQuireBio™ QP13496100ug
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
A cDNA sequence encoding the Shiga Like Toxin 2 was constructed and used to recombinantly synthesize the protein.
Especificaciones
Nombre Shiga Like Toxin 2 Protein
Cantidad 100 μg
Estado normativo Research Use Only
Concentración de endotoxinas Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Tipo de producto Recombinant Protein
Reactividad cruzada Other
Especie E. coli
Etiqueta de proteína His
Tampón Shiga Like Toxin 2 protein solution contains PBS, 50mM arginine and 0.05% sodium azide.
Grado de pureza o calidad Protein is >95% pure as determined by 12% PAGE (coomassie staining).
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.