Learn More
Gibco™ Human IL-15 Recombinant Protein
Código de producto. 10303723
Change view
Click to view available options
Cantidad:
1000 μg
100 μg
10 μg
Tamaño de la unidad:
1 miligramo
cada uno
Código de producto. Cantidad unitSize
10303723 10 μg cada uno
10013534 100 μg cada uno
11632069 1000 μg 1 miligramo
3 options
Código de producto. 10303723
Marca: Gibco™ PHC9154
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Recombinant Protein
Carrier-Free Reconstitution: We recommend reconstituting the lyophilized protein in sterile double distilled water containing at least 0.1% human serum albumin or bovine serum albumin to prepare a stock solution of =1 mg/mL of the protein. The reconstituted protein should be apportioned into working aliquots and stored at =-20°C. Storage: The lyophilized protein is stable for a few weeks at room temperature, but is best stored at -20°C. Reconstituted human IL-15 should be stored at =-20°C (not in a frost-free freezer). Keep freeze-thaw cycles to a minimum.
IL-15 (Interleukin 15 ) is a cytokine that regulates T and natural killer cell activation and proliferation. IL-15 and IL-2 share many biological activities as both have been found to bind common hematopoietin receptor subunits, and may compete for the same receptor, and thus negatively regulate each other's activity. The number of CD8+ memory cells is shown to be controlled by a balance between IL-15 and IL-2. IL-15 induces the activation of JAK kinases, as well as the phosphorylation and activation of transcription activators STAT3, STAT5, and STAT6. In mouse, studies suggest that IL-15 may increase the expression of apoptosis inhibitor Bcl-xL, possibly through the transcription activation activity of STAT6, and thus prevent apoptosis. IL-15 plays an important role in the growth and differentiation of T and B lymphocytes, natural killer cells, macrophages, and monocytes as well as activation of a number of important intracellular signaling molecules. As such, IL-15 could be essential for the immune responses, allograft rejection, and the pathogenesis of autoimmune diseases. Further, IL-15 is a widely expressed pro-inflammatory cytokine and has been shown to play a role in several inflammatory disorders, including rheumatoid arthritis, psoriasis and pulmonary inflammatory diseases. Emerging data suggest that IL-15 may serve as a good therapeutic target, as there appears to be a beneficial effect of IL-15 neutralization in models of psoriasis and diabetes.
TRUSTED_SUSTAINABILITY
Especificaciones
Número de acceso P40933
Para utilizar con (aplicación) Bioactivity
Formulación Protein with no preservative
ID de gen (Entrez) 3600
Peso molecular 12.9 kDa
Nombre Human IL-15
Método de purificación Purified
Cantidad 10 μg
Requisitos de almacenamiento -20°C
Estado normativo RUO
Concentración de endotoxinas <1 EU/mL
Alias de gen AI503618; il 15; Il15; IL-15; ILN; Interleukin; interleukin 15; Interleukin15; interleukin-15; MGC9721; M-IL-15
Nombre común IL-15
Símbolo de gen IL15
Actividad biológica ED50 ≤ 0.5 ng/mL; determined by the dose-dependent proliferation of CTLL-2 cells.
Tipo de producto Protein
Conjugado Unconjugated
Especie Human
Recombinante Recombinant
Secuencia Human IL-15 recombinant protein contains 115 amino acids
Contenido y almacenamiento -20°C
Clasificación Carrier-Free
Actividad ED50 ≤ 0.5 ng/mL; determined by the dose-dependent proliferation of CTLL-2 cells.
Nivel de endotoxinas <1 EU/ml
Condiciones de envío Approved for shipment on Wet or Dry Ice
Sistema de expresión E. coli
Familia de proteínas Cytokines & Receptors
Forma de proteína Recombinant, Ligand
Formulario Lyophilized
Subtipo de proteína Interleukins
Categoría de investigación Cell Cycle & Proliferation, Oncology, Immunology, Signal Transduction
Línea de productos Gibco
Grado de pureza o calidad ≥98% by SDS-PAGE