Learn More
VGLUT2 Antibody (CL2921) [Alexa Fluor™ 750], Novus Biologicals™
Código de producto. 30303351 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
30303351 0.1 mL 0.10 ml
1 options
Código de producto. 30303351 Proveedor Novus Biologicals N.º de proveedor NBP246640AF750
Mouse Monoclonal Antibody
VGLUT2 Monoclonal antibody specifically detects VGLUT2 in Human,Mouse,Rat samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Especificaciones
Antígeno VGLUT2
Aplicaciones Immunohistochemistry, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon CL2921
Conjugado Alexa Fluor 750
Formulación 50mM Sodium Borate
Alias de gen differentiation-associated BNPI, differentiation-associated Na(+)-dependent inorganic phosphate cotransporter, differentiation-associated Na-dependent inorganic phosphate cotransporter, DNPI, solute carrier family 17 (sodium-dependent inorganic phosphate cotransporter), member 6, solute carrier family 17 member 6, vesicular glutamate transporter 2, VGLUT2
Especie del huésped Mouse
Inmunógeno This antibody was developed using a recombinant protein derived from Q9P2U8, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Neuroscience
Primario o secundario Primary
ID de gen (Entrez) 57084
Especies diana Human, Mouse, Rat
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG1