missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
VEGF Monoclonal antibody specifically detects VEGF in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antígeno | VEGF |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Monoclonal |
| Clon | VEGFA/7636R |
| Conjugado | Alexa Fluor 405 |
| Formulación | 50 mM Sodium Borate |
| Alias de gen | MVCD1, vascular endothelial growth factor A, Vascular permeability factor, VEGF-A, VEGFMGC70609, VPFvascular endothelial growth factor |
| Especie del huésped | Rabbit |
| Inmunógeno | Recombinant fragment (around aa1-200) of human VEGF protein (exact sequence is proprietary) |
| Método de purificación | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?