missing translation for 'onlineSavingsMsg'
Learn More
Learn More
VCP Polyclonal Antibody, OriGene
Rabbit Polyclonal Antibody
Marca: Origene Technologies TA890028
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The protein encoded by this gene is a member of a family that includes putative ATP-binding proteins involved in vesicle transport and fusion, 26S proteasome function, and assembly of peroxisomes. This protein, as a structural protein, is associated with clathrin, and heat-shock protein Hsc70, to form a complex. It has been implicated in a number of cellular events that are regulated during mitosis, including homotypic membrane fusion, spindle pole body function, and ubiquitin-dependent protein degradation.Especificaciones
| VCP | |
| Polyclonal | |
| PBS with 50% glycerol and 0.02% sodium azide | |
| P46462, P55072, Q01853 | |
| Vcp | |
| Recombit protein of human VCP | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| VCP | |
| CDC48, TERA, p97 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 116643, 269523, 7415 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?