missing translation for 'onlineSavingsMsg'
Learn More

Vanilloid R1/TRPV1 Antibody, Novus Biologicals™

Código de producto. 18264990 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.025 mL
0.1 mL
Tamaño de la unidad:
0.10 ml
25 microlitros
Código de producto. Cantidad unitSize
18264990 0.1 mL 0.10 ml
18204761 0.025 mL 25 microlitros
2 options
Este artículo no se puede devolver. Vea la política de devoluciones

Código de producto. 18264990

Marca: Novus Biologicals NB10098886

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Este artículo no se puede devolver. Vea la política de devoluciones

Rabbit Polyclonal Antibody has been used in 5 publications

Vanilloid R1/TRPV1 Polyclonal specifically detects Vanilloid R1/TRPV1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY

Especificaciones

Antígeno Vanilloid R1/TRPV1
Aplicaciones Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Clasificación Polyclonal
Concentración LYOPH
Conjugado Unconjugated
Dilución Western Blot 1:1000, Immunohistochemistry 1:1000, Immunocytochemistry/Immunofluorescence 1:10-1:500, Immunohistochemistry-Paraffin 1:1000
Formulación Lyophilized from whole antisera with No Preservative
N.º de referencia del gen Q8NER1
Alias de gen Capsaicin receptor, Osm-9-like TRP channel 1, OTRPC1, transient receptor potential cation channel subfamily V member 1, transient receptor potential cation channel, subfamily V, member 1, transient receptor potential vanilloid 1a, transient receptor potential vanilloid 1b, TrpV1, Vanilloid receptor 1, vanilloid receptor subtype 1, VR1, VR1DKFZp434K0220
Símbolos de los genes TRPV1
Especie del huésped Rabbit
Inmunógeno A synthetic peptide from the 1t cytoplasmic loop of human Vanilloid R1/TRPV1 conjugated to blue carrier protein was used as the antigen.
Método de purificación Unpurified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Neuroscience, Sensory Systems
Primario o secundario Primary
ID de gen (Entrez) 7442
Reconstitución Reconstitute in 0.1 ml of sterile water. Centrifuge to remove any insoluble material. Glycerol may be added (1:1) for additional stability. Please note the sample size is provided in reconstituted format.
Especies diana Human, Mouse
Contenido y almacenamiento Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Formulario Antisera
Isotype IgG
Mostrar más Mostrar menos

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.