missing translation for 'onlineSavingsMsg'
Learn More

USP30 Antibody (CL4438) [Janelia Fluor™ 549], Novus Biologicals™

Código de producto. 30298329 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Código de producto. Cantidad unitSize
30298329 0.1 mL 0.10 ml
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Este artículo no se puede devolver. Vea la política de devoluciones
Código de producto. 30298329 Proveedor Novus Biologicals N.º de proveedor NBP259049JF549

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Mouse Monoclonal Antibody

USP30 Monoclonal antibody specifically detects USP30 in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antígeno USP30
Aplicaciones Immunohistochemistry, Immunohistochemistry (Paraffin)
Clasificación Monoclonal
Clon CL4438
Conjugado Janelia Fluor 549
Formulación 50mM Sodium Borate
Alias de gen Deubiquitinating enzyme 30, EC 3.1.2.15, EC 3.4.19.12, FLJ40511, MGC10702, ubiquitin carboxyl-terminal hydrolase 30, ubiquitin specific peptidase 30, ubiquitin specific protease 30, ubiquitin thioesterase 30, Ubiquitin thiolesterase 30, ubiquitin-specific protease 30, Ubiquitin-specific-processing protease 30, Ub-specific protease 30
Especie del huésped Mouse
Inmunógeno This antibody was developed using a recombinant protein derived from Q70CQ3, with the exact immunogen sequence remaining proprietary.
Método de purificación Protein A purified
Cantidad 0.1 mL
Estado normativo RUO
Disciplina de investigación Cancer, Cell Biology, Endocrinology, Signal Transduction
Primario o secundario Primary
ID de gen (Entrez) 84749
Especies diana Human
Contenido y almacenamiento Store at 4⪚C in the dark.
Formulario Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.