missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ uPAR Antibody (ATN-658) - Humanized, Novus Biologicals™
Tienda Bio Techne ProductosDescription
uPAR Monoclonal antibody specifically detects uPAR in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
Specifications
| Antígeno | uPAR |
| Aplicaciones | Flow Cytometry, ELISA, Functional Assay |
| Clasificación | Monoclonal |
| Clon | ATN-658 |
| Conjugado | Unconjugated |
| Formulación | Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2) |
| N.º de referencia del gen | Q03405 |
| Alias de gen | CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3 |
| Especie del huésped | Mouse |
| Inmunógeno | PLAUR/ uPAR/ CD87 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?