missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
UGT2B10 Polyclonal antibody specifically detects UGT2B10 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antígeno | UGT2B10 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:500 - 1:2000 |
| Formulación | PBS (pH 7.3), 50% glycerol |
| Alias de gen | EC 2.4.1.17, MGC142209, UDP glucuronosyltransferase 2 family, polypeptide B10, UDP glucuronosyltransferase 2B10, UDP glycosyltransferase 2 family, polypeptide B10, UDP-glucuronosyltransferase 2B10, UDPGT 2B10, UDPGT2B10 |
| Especie del huésped | Rabbit |
| Inmunógeno | Recombinant fusion protein containing a sequence corresponding to amino acids 20-280 of human UGT2B10 (NP_001066.1). GSCGKVLVWAAEYSLWMNMKTILKELVQRGHEVTVLASSASILFDPNDSSTLKLEVYPTSLTKTEFENIIMQLVKRLSEIQKDTFWLPFSQEQEILWAINDIIRNFCKDVVSNKKLMKKLQESRFDIVFADAYLPCGELLAELFNIPFVYSHSFSPGYSFERHSGGFIFPPSYVPVVMSKLSDQMTFMERVKNMLYVLYFDFWFQIFNMKKWDQFYSEVLGRPTTLSETMRKADIWLMRNSWNFKFPHPFLPNVDFVGGLH |
| Método de purificación | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?