Learn More
Tyrosine Hydroxylase Antibody, Novus Biologicals™
Código de producto. 18757973 Tienda Bio Techne Productos
Change view
Click to view available options
Cantidad:
0.1 mL
Tamaño de la unidad:
0.10 ml
18757973 0.1 mL 0.10 ml
1 options
Código de producto. 18757973
Marca: Novus Biologicals NB300109
Rabbit Polyclonal Antibody has been used in 147 publications
Tyrosine Hydroxylase Polyclonal specifically detects Tyrosine Hydroxylase in Human, Mouse, Rat, Drosophila, Insect, Mammal samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating, Immunohistochemistry Whole-Mount, Knockout Validated, Dual RNAscope ISH-IHC.
TRUSTED_SUSTAINABILITY
Especificaciones
Cantidad 0.1 mL
Estado normativo RUO
For Research Use Only