missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ TXNL2 Recombinant Protein
Descripción
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Specifications
| Número de acceso | AAH05289 |
| Para utilizar con (aplicación) | Antibody Production, Protein Array, ELISA, Western Blot |
| Formulación | 50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer |
| ID de gen (Entrez) | 10539 |
| Peso molecular | 62.59 |
| Nombre | GLRX3 (Human) Recombinant Protein (P01) |
| Intervalo de pH | 8 |
| Método de preparación | In vitro wheat germ expression system |
| Método de purificación | Glutathione Sepharose 4 Fast Flow |
| Pruebas de control de calidad | 12.5% SDS-PAGE Stained with Coomassie Blue. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?