missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Epredia™ Lab Vision™ Tubulin-α, Rabbit Polyclonal Antibody
Provide accurate, reproducible results with the Epredia™ Tubulin-α, Rabbit Polyclonal Antibody.
132.60€ - 550.80€
Especificaciones
Antígeno | Tubulin-alpha |
---|---|
Aplicaciones | Western Blot |
Clasificación | Polyclonal |
Conjugado | Unconjugated |
Especie del huésped | Rabbit |
Descripción
Microtubules, the major cytoskeletal elements found in all eukaryotic cells, consist of Tubulin, which is a dimer of two 55kDa subunits: α and β. Microtubules play key roles in chromosome segregation in mitosis, intracellular transport, ciliary and flagellar bending, and structural support of the cytoskeleton.Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Cow, Pig, Rat, Mouse, Guinea Pig, Gerbil, Frog and Chicken.
Epitope: aa 426-450
Immunogen: A synthetic peptide derived from aa 426-450 of human tubulin-α
Molecular Weight: 57kDa
Positive Control: MAD109 cells, MCF-7 cells, skin or lung
Cellular Localization: Cytoplasmic This antibody is excellent for staining of formalin-fixed, paraffin-embedded tissues.
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
- Western Blotting
Especificaciones
Tubulin-alpha | |
Polyclonal | |
Rabbit | |
Cytoskeletal Protein | |
A synthetic peptide derived from aa 426-450 of human tubulin alpha | |
Primary |
Western Blot | |
Unconjugated | |
IVD | |
Bovine, Chicken, Frog, Gerbil, Guinea Pig, Human, Mouse, Porcine | |
IgG |