missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Descripción
TTC14 Polyclonal antibody specifically detects TTC14 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Especificaciones
Especificaciones
| Antígeno | TTC14 |
| Aplicaciones | Immunohistochemistry (Paraffin) |
| Clasificación | Polyclonal |
| Conjugado | mFluor Violet 500 SE |
| Dilución | Immunohistochemistry-Paraffin |
| Formulación | 50mM Sodium Borate |
| Alias de gen | DKFZp313M1015, DRDL5813, KIAA1980FLJ00166, PRO19630, tetratricopeptide repeat domain 14, tetratricopeptide repeat protein 14, TPR repeat protein 14 |
| Especie del huésped | Rabbit |
| Inmunógeno | Produced in rabbits immunized with E. coli-derived Human TTC14 fragment. |
| Método de purificación | Antigen and protein A Affinity-purified |
| Mostrar más |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?