missing translation for 'onlineSavingsMsg'
Learn More

TSH-Receptor, A-Chain Mouse, Clone: SPM223, enQuireBio™

Mouse Monoclonal Antibody

199.00€ - 554.40€

Especificaciones

Antígeno TSH-Receptor, A-Chain
Clon SPM223
Concentración 200μg/mL
Aplicaciones Immunohistochemistry
Clasificación Monoclonal
Ver más especificaciones

Productos 2
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
Código de producto Marca Cantidad Precio Cantidad y disponibilidad  
15901028
Ver documentos
EnquireBio
7253-MSM1X-P0
20 μg Este artículo actualmente no está disponible o ha sido discontinuado.
Ver la página del producto para posibles alternativas
N/A View alternative products
missing translation for 'toSaveUpto' 
»
15911028
missing translation for 'viewDocuments'
EnquireBio
7253-MSM1X-P1
100 μg
554.40€
100 microgramos
View alternative products
missing translation for 'toSaveUpto' 
»
Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.
Description

Description

Specifications

Specifications

TSH-Receptor, A-Chain
200μg/mL
Monoclonal
Mouse
10mM PBS with 0.05% BSA and 0.05% Azide
7253
Recombinant human TSHR, A-Chain protein
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
62kDa
SPM223
Immunohistochemistry
Unconjugated
RUO
XP_005268094
TSHR
IgG1
Protein A/G Purified
Cytoplasmic
293T Cells. Thyroid gland
Videos
SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.