Learn More
TSG101 Antibody, Allophycocyanin, Novus Biologicals™
Código de producto. 18641866 Tienda Bio Techne Productos
Código de producto. 18641866 Proveedor Novus Biologicals N.º de proveedor NBP298994APC
Rabbit Polyclonal Antibody
TSG101 Polyclonal antibody specifically detects TSG101 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunoprecipitation