missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TSG-6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 13321-1-AP-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
TSG6 is a secretory protein that contains a hyaluro-binding domain, and thus is a member of the hyaluro-binding protein family. The hyaluro-binding domain is involved in extracellular matrix stability and cell migration. This protein forms a stable complex with inter-alpha-inhibitor (I alpha I), and thus enhances the serine protease inhibitory activity of I alpha I, which is important in the protease network associated with inflammation. The expression of this gene can be induced by tumor necrosis factor alpha and interleukin-1. The expression can also be induced by mechanical stimuli in vascular smooth muscle cells, and is correlated with proteoglycan synthesis and aggregation.Especificaciones
| TSG-6 | |
| Polyclonal | |
| Unconjugated | |
| TNFAIP6 | |
| Hyaluronate binding protein, TNF alpha induced protein 6, TNF stimulated gene 6 protein, TNFAIP6, TSG 6, TSG6 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 21930, 7130, 84397 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.23 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O08859, P98066 | |
| TNFAIP6 | |
| TSG-6 Fusion Protein Ag3885 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?