missing translation for 'onlineSavingsMsg'
Learn More

TSG-6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Código de producto. p-7204645 Shop All Proteintech Products
Change view
Click to view available options
Cantidad:
150 μL
20 μL
Unit Size:
150 microlitros
20 microlitros
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Cantidad unitSize
16821474 20 μL 20 microlitros
16811474 150 μL 150 microlitros
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16821474 Supplier Proteintech Supplier No. 133211AP20UL

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TSG6 is a secretory protein that contains a hyaluro-binding domain, and thus is a member of the hyaluro-binding protein family. The hyaluro-binding domain is involved in extracellular matrix stability and cell migration. This protein forms a stable complex with inter-alpha-inhibitor (I alpha I), and thus enhances the serine protease inhibitory activity of I alpha I, which is important in the protease network associated with inflammation. The expression of this gene can be induced by tumor necrosis factor alpha and interleukin-1. The expression can also be induced by mechanical stimuli in vascular smooth muscle cells, and is correlated with proteoglycan synthesis and aggregation.

Specifications

Antígeno TSG-6
Aplicaciones Western Blot
Clasificación Polyclonal
Concentración 0.23 mg/mL
Conjugado Unconjugated
Formulación PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
génica TNFAIP6
N.º de referencia del gen O08859, P98066
Alias de gen Hyaluronate binding protein, TNF alpha induced protein 6, TNF stimulated gene 6 protein, TNFAIP6, TSG 6, TSG6
Símbolos de los genes TNFAIP6
Especie del huésped Rabbit
Inmunógeno TSG-6 Fusion Protein Ag3885
Método de purificación Antigen Affinity Chromatography
Cantidad 20 μL
Estado normativo RUO
Primario o secundario Primary
ID de gen (Entrez) 21930, 7130, 84397
Especies diana Human, Rat, Mouse
Contenido y almacenamiento -20°C
Tipo de producto Antibody
Formulario Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.