missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TSEN54 Polyclonal antibody specifically detects TSEN54 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
Specifications
| Antígeno | TSEN54 |
| Aplicaciones | Western Blot |
| Clasificación | Polyclonal |
| Conjugado | Unconjugated |
| Dilución | Western Blot 1:500-1:2000 |
| Formulación | PBS (pH 7.3), 50% glycerol |
| Alias de gen | hsSen54, PCH2A, PCH4, sen54, SEN54 homolog, SEN54FLJ37147, SEN54L, tRNA splicing endonuclease 54 homolog (S. cerevisiae), tRNA splicing endonuclease 54 homolog (SEN54, S. cerevisiae), tRNA-intron endonuclease Sen54, tRNA-splicing endonuclease subunit Sen54 |
| Especie del huésped | Rabbit |
| Inmunógeno | Recombinant fusion protein containing a sequence corresponding to amino acids 1-300 of human TSEN54 (NP_997229.2). MEPEPEPAAVEVPAGRVLSARELFAARSRSQKLPQRSHGPKDFLPDGSAAQAERLRRCREELWQLLAEQRVERLGSLVAAEWRPEEGFVELKSPAGKFWQTMGFSEQGRQRLHPEEALYLLECGSIHLFHQDLPLSIQEAYQLLLTDHTVTFLQYQVFSHLKRLGYVVRRFQPSSVLSPYERQLNLDASVQHLEDGDGKRKRSSSSPRSINKKAKALDNSLQPKSLAASSPPPCSQPSQCPEEKPQESSPMKGPGGPFQLLGSLGPSPGPAREGVGCSWESGRAENGVTGAGKRRWNFEQ |
| Método de purificación | Affinity purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?