Learn More
TRPV2 Mouse, anti-Human, Clone: 2F11H3, Proteintech
Mouse Monoclonal Antibody
Marca: Proteintech 68563-1-IG-20UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The TRPV2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human TRPV2. This antibody recognizes human, mouse, rat antigen. The TRPV2 antibody has been validated for the following applications: WB, ELISA analysis.
Especificaciones
| TRPV2 | |
| 2F11H3 | |
| Osm 9 like TRP channel 2, OTRPC2, TRPV2, VRL, VRL 1, VRL1 | |
| 20 μL | |
| 51393 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG2a |